ContactUsLinkComponent
Neulog Sense C Language Coding Unit
Catalogue # 470232-640
Fournisseur:  Wards
undefined
Neulog Sense C Language Coding Unit
Catalogue # 470232-640
Fournisseur:  Wards
Supplier Number:  CARM202
Spécifications
  • Description:
    Neulog Sense C Language Coding Unit
  • Cat. no.:
    470232-640
Spécifications
À propos de cet article
The Sense C is a trainer that allows a user to study and exercise C language and to program Neulog Sense in C language.
  • Sense is a STEM computer programing and engineering learning platform
A student from the age of 8 years old can begin programming sense via visual block programming (free for download) and then solve any challenge imaginable by plugging various sensors, coding modules, or robotic parts into sense.
Once a student grasps the fundamentals of visual programing they can then purchase modules to program in a variety of other languages such as C and Python. Sense will provide STEM and coding education from ages 8 – 21.
C is the coding language for creating machine programs and is the most needed language in the industry right now. Students can use the Sense C language programming unit to train on how to code in C an then program there Sense bots and Neulog sensors to solve real world STEM problems.