ContactUsLinkComponent
Tricorn™ Beakers, Triangular, Simport Scientific
Catalogue # CA73960-144
Fournisseur:  Simport Scientific
Fournisseur #: B700-800
undefined
Tricorn™ Beakers, Triangular, Simport Scientific
Catalogue # CA73960-144
Fournisseur:  Simport Scientific
Supplier Number:  B700-800
Fournisseur #: B700-800
S'identifier pour la disponibilité
Select an Option
107 mm
Select an Option
50 ml
Select an Option
800 ml
Select an Option
133 mm
Select an Option
CA73960-144
Select an Option
B700-800
À propos de cet article
These Tricorn™ beakers are made of polypropylene, provide three dripless pouring spouts that offers more security.
  • Autoclavable
  • Tough and unbreakable
Suitable for use with commonly used acids, alkalis and solvents.
Packaging: All sizes are available 100 per case.
Frequently Bought Together