Semi-Micro Cuvettes, Fireflysci
Marketsource
Supplier: FIREFLYSCI, INC.
Jump to:
À propos de cet article
Standard Semi-micro spectrophotometer cuvette with PTFE Cover.
- Gives highly accurate results, only need to test sample once
- Comes in several different materials: Optical Glass, UV Quartz, IR Quartz
- Excellent transmission values
- Fire-fused to ensure that cuvettes are robust and durable
Delivery information: Lightpaths from 5 – 100 mm are available.
7 Options Available Below
Product Details & Documents
Standard Semi-micro spectrophotometer cuvette with PTFE Cover.
- Gives highly accurate results, only need to test sample once
- Comes in several different materials: Optical Glass, UV Quartz, IR Quartz
- Excellent transmission values
- Fire-fused to ensure that cuvettes are robust and durable
Delivery information: Lightpaths from 5 – 100 mm are available.
Product Family Options
Informations sur le produit
- DésignationCapacitéOuter HeightOuter WidthLightpathCover TypeAvailabilityPrice
- Supplier Number: 9UV5
- Cuvette, Outer Length: 12.5 mm1.400 ml45 mm12.5 mm10 mmPTFELog in pour la disponibilitéEach$562.18Supplier Number: 9UV10
- Cuvette, Outer Length: 22.5 mm2.800 ml45 mm12.5 mm20 mmPTFELog in pour la disponibilitéEach$615.71Supplier Number: 9UV20Spécifications:MarketsourceMarketsource
- Cuvette, Outer Length: 32.5 mm4.200 ml45 mm12.5 mm30 mmPTFELog in pour la disponibilitéEach$665.95Supplier Number: 9UV30Spécifications:MarketsourceMarketsource
- Cuvette, Outer Length: 42.5 mm5.600 ml45 mm12.5 mm40 mmPTFELog in pour la disponibilitéEach$795.74Supplier Number: 9UV40Spécifications:MarketsourceMarketsource
- Cuvette, Outer Length: 52.5 mm7.000 ml45 mm12.5 mm50 mmPTFELog in pour la disponibilitéEach$879.62Supplier Number: 9UV50Spécifications:MarketsourceMarketsource
- Cuvette, Outer Length: 102.5 mm14.000 ml45 mm12.5 mm100 mmPTFELog in pour la disponibilitéEach$1,195.33Supplier Number: 9UV100Spécifications:MarketsourceMarketsource