LB Agar Plates with 30 µg/mL Kanamycin, X-gal and IPTG, Teknova
Catalogue # CA100217-644
Fournisseur: Teknova
Numéro CAS:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Description:LB Agar with 0.1 mM IPTG, 60 µg/mL X-gal
- Size:100 x 100 mm
- Environmentally Preferable:
- Cat. no.:CA100217-644
- Supplier No.:L3908
Spécifications
À propos de cet article
Ingredients: 1.0% tryptone, 0.5% yeast extract, 1.0% sodium chloride (NaCl), 1.5% agar (15.0 g/L), 30 µg/mL kanamycin.
- Sleeve of 20 plates
- Sterile
Ambient shipping conditions. Refrigerate to store.