Standard For Osmometer, Advanced Instruments | Avantor
Standard For Osmometer, Advanced Instruments
Standard For Osmometer, Advanced Instruments
Certificates
Standard For Osmometer, Advanced Instruments
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Calibration standard is used to check the perforance of osmometer and test methods.
Documents
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • 200 mOsm Calibration Standard
    10×2 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $402.56
    $402.56
    Pack of 10
       $402.56
    Supplier Number: 3MA020
  • 400 mOsm Calibration Standard
    10×2 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $402.56
    $402.56
    Pack of 10
       $402.56
    Supplier Number: 3MA040
  • 1000 mOsm Calibration Standard
    10×2 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $402.56
    $402.56
    Pack of 10
       $402.56
    Supplier Number: 3MA100