Microscope slides, Kemtech America
Supplier: Kemtech America
Jump to:
About this item
Kemtech America slides are in compliance with iso 8037/1 international standard in dimension. each slide is pre cleaned and thouroughly examined to ensure premium quality
- Super grade and super white glass available
- 45 degree clipped and 90 degree corners available
- color frosted
- Iso 8037/1 compiant
Certifications: ISO Certified: GB/T 19001-2008/ISO 9001:2008
41 Options Available Below
Product Details & Documents
Kemtech America slides are in compliance with iso 8037/1 international standard in dimension. each slide is pre cleaned and thouroughly examined to ensure premium quality
- Super grade and super white glass available
- 45 degree clipped and 90 degree corners available
- color frosted
- Iso 8037/1 compiant
Certifications: ISO Certified: GB/T 19001-2008/ISO 9001:2008
Product Family Options
Product Information
- DésignationColorÉpaisseurL×lAvailabilityPrice
- Slides, super grade, ground edges, plain, 45° clipped cornersPlain1.1 mm75×25 mmDiscontinued-Supplier Number: 0302-0022Specifications:
More
Specifications
Cat. No.76203-940 - Slides, super grade, ground edges, single frosted one end, 45° clipped cornersPlain1.1 mm75×25 mmLog in for availabilityCase of 1,440$311.71Supplier Number: 0302-2122Specifications:
More
Specifications
Cat. No.76203-942 - Slides, super grade, ground edges, double-frosted one end, 45° clipped cornersPlain1.1 mm75×25 mmDiscontinued-Supplier Number: -Specifications:
More
Specifications
Cat. No.76203-944 - Slides, super grade, ground edges, colored single frosted one end, 45° clipped corners, printableBlue1.1 mm75×25 mmLog in for availabilityCase of 1,440$391.21Supplier Number: 0302-7121-02Specifications:
More
Specifications
Cat. No.76203-946 - Slides, super grade, ground edges, colored single frosted one end, 45° clipped corners, printableGreen1.1 mm75×25 mmLog in for availabilityCase of 1,440$391.21Supplier Number: 0302-7121-08Specifications:
More
Specifications
Cat. No.76203-948 - Slides, super grade, ground edges, coloredsingle frosted one end, 45° clipped corners, printablePurple1.1 mm75×25 mmLog in for availabilityCase of 1,440$391.21Supplier Number: 0302-7121-09Specifications:
More
Specifications
Cat. No.76204-146 - Slides, super grade, ground edges, colored single frosted one end, 45° clipped corners, printablePink1.1 mm75×25 mmLog in for availabilityCase of 1,440$391.21Supplier Number: 0302-7121-12Specifications:
More
Specifications
Cat. No.76204-148 - Slides, super grade, ground edges, double-frosted one end, 90° cornersPlain1.1 mm75×25 mmLog in for availabilityCase of 1,440$339.65Supplier Number: 0303-2222Specifications:
More
Specifications
Cat. No.76204-150 - Slides, super grade, ground edges, colored single frosted one end, 90° corners, printableBlue1.1 mm75×25 mmLog in for availabilityCase of 1,440$391.21Supplier Number: 0303-7121-02Specifications:
More
Specifications
Cat. No.76204-152 - Slides, super grade, ground edges, colored single frosted one end, 90° corners, printableGreen1.1 mm75×25 mmLog in for availabilityCase of 1,440$391.21Supplier Number: 0303-7121-08Specifications:
More
Specifications
Cat. No.76204-154 - Slides, super grade, ground edges, colored single frosted one end, 90° corners, printablePurple1.1 mm75×25 mmLog in for availabilityCase of 1,440$391.21Supplier Number: 0303-7121-09Specifications:
More
Specifications
Cat. No.76204-156 - Slides, super grade, ground edges, colored single frosted one end, 90° corners, printablePink1.1 mm75×25 mmLog in for availabilityCase of 1,440$391.21Supplier Number: 0303-7121-12Specifications:
More
Specifications
Cat. No.76204-158 - Slides, super grade, ground edges, colored single frosted one end, 90° corners, printableWhite1.1 mm75×25 mmLog in for availabilityCase of 1,440$391.21Supplier Number: 0303-7121-16Specifications:
More
Specifications
Cat. No.76204-160 - Slides, super grade, ground edges, colored single frosted one end, 90° corners, printableYellow1.1 mm75×25 mmLog in for availabilityCase of 1,440$391.21Supplier Number: 0303-7121-17Specifications:
More
Specifications
Cat. No.76204-162 - Slides, super grade, beveled edges, plain, 45° clipped cornersPlain1.1 mm75×25 mmLog in for availabilityCase of 1,440$348.29Supplier Number: 0304-0022Specifications:
More
Specifications
Cat. No.76204-164 - Slides, super grade, beveled edges, single frosted one end, 45° clipped cornersPlain1.1 mm75×25 mmLog in for availabilityCase of 1,440$354.72Supplier Number: 0304-2122Specifications:
More
Specifications
Cat. No.76204-166 - Slides, super grade, beveled edges, double-frosted one end, 45° clipped cornersPlain1.1 mm75×25 mmLog in for availabilityCase of 1,440$361.17Supplier Number: 0304-2222Specifications:
More
Specifications
Cat. No.76204-168 - Slides, super grade, beveled edges, colored single frosted one end, 45° clipped corners, printableBlue1.1 mm75×25 mmLog in for availabilityCase of 1,440$404.16Supplier Number: 0304-7121-02Specifications:
More
Specifications
Cat. No.76204-170 - Slides, super grade, beveled edges, colored single frosted one end, 45° clipped corners, printableGreen1.1 mm75×25 mmLog in for availabilityCase of 1,440$404.16Supplier Number: 0304-7121-08Specifications:
More
Specifications
Cat. No.76204-172 - Slides, super grade, beveled edges, coloredsingle frosted one end, 45° clipped corners, printablePurple1.1 mm75×25 mmLog in for availabilityCase of 1,440$404.16Supplier Number: 0304-7121-09Specifications:
More
Specifications
Cat. No.76204-174 - Slides, super grade, beveled edges, colored single frosted one end, 45° clipped corners, printablePink1.1 mm75×25 mmLog in for availabilityCase of 1,440$404.16Supplier Number: 0304-7121-12Specifications:
More
Specifications
Cat. No.76204-176 - Slides, super grade, beveled edges, colored single frosted one end, 45° clipped corners, printableWhite1.1 mm75×25 mmDiscontinued-Supplier Number: -Specifications:
More
Specifications
Cat. No.76204-178 - Slides, super grade, beveled edges, colored single frosted one end, 45° clipped corners, printableYellow1.1 mm75×25 mmLog in for availabilityCase of 1,440$404.16Supplier Number: 0304-7121-17Specifications:
More
Specifications
Cat. No.76204-180 - Slides, super colored glass, ground edges, plain, 45°clipped cornersPlain1.1 mm75×25 mmLog in for availabilityCase of 1,440$472.94Supplier Number: 0312-0021Specifications:
More
Specifications
Cat. No.76204-182 - Slides, super colored glass, ground edges, single frosted one end, 45°clipped cornersPlain1.1 mm75×25 mmLog in for availabilityCase of 1,440$483.75Supplier Number: 0312-2121Specifications:
More
Specifications
Cat. No.76204-184 - Slides, super colored glass, ground edges, double-frosted one end, 45°clipped cornersPlain1.1 mm75×25 mmLog in for availabilityCase of 1,440$483.75Supplier Number: 0312-2221Specifications:
More
Specifications
Cat. No.76204-186 - Slides, super colored glass, ground edges, plain, 90° cornersPlain1.1 mm75×25 mmLog in for availabilityCase of 1,440$472.94Supplier Number: 0313-0021Specifications:
More
Specifications
Cat. No.76204-188 - Slides, super colored glass, ground edges, single frosted one end, 90° cornersPlain1.1 mm75×25 mmLog in for availabilityCase of 1,440$483.75Supplier Number: 0313-2121Specifications:
More
Specifications
Cat. No.76204-190 - Slides, super colored glass, ground edges, double-frosted one end, 90° cornersPlain1.1 mm75×25 mmLog in for availabilityCase of 1,440$483.75Supplier Number: 0313-2221Specifications:
More
Specifications
Cat. No.76204-192 - Slides, super colored glass, ground edges, colored single frosted one end, 90° corners, printableBlue1.1 mm75×25 mmLog in for availabilityCase of 1,440$485.83Supplier Number: 0313-7121-02Specifications:
More
Specifications
Cat. No.76204-194 - Slides, super colored glass, ground edges, colored single frosted one end, 90° corners, printableGreen1.1 mm75×25 mmLog in for availabilityCase of 1,440$485.83Supplier Number: 0313-7121-08Specifications:
More
Specifications
Cat. No.76204-196 - Slides, super colored glass, ground edges, coloredsingle frosted one end, 90° corners, printablePurple1.1 mm75×25 mmLog in for availabilityCase of 1,440$485.83Supplier Number: 0313-7121-09Specifications:
More
Specifications
Cat. No.76204-198 - Slides, super colored glass, ground edges, colored single frosted one end, 90° corners, printablePink1.1 mm75×25 mmLog in for availabilityCase of 1,440$485.83Supplier Number: 0313-7121-12Specifications:
More
Specifications
Cat. No.76204-200 - Slides, super colored glass, ground edges, colored single frosted one end, 90° corners, printableWhite1.1 mm75×25 mmLog in for availabilityCase of 1,440$485.83Supplier Number: 0313-7121-16Specifications:
More
Specifications
Cat. No.76204-202 - Slides, super colored glass, ground edges, colored single frosted one end, 90° corners, printableYellow1.1 mm75×25 mmLog in for availabilityCase of 1,440$485.83Supplier Number: 0313-7121-17Specifications:
More
Specifications
Cat. No.76204-204 - Discontinued-Supplier Number: -Specifications:
More
Specifications
Cat. No.76204-148 - Discontinued-Supplier Number: -Specifications:
More
Specifications
Cat. No.76204-146 - Slides, super colored glass, beveled edges, colored single frosted one end, 45° clipped corners, printableBlue1.1 mm75×25 mmLog in for availabilityCase of 1,440$520.25Supplier Number: 0314-7121-02Specifications:
More
Specifications
Cat. No.76204-210 - Slides, super colored glass, beveled edges, colored single frosted one end, 45° clipped corners, printableGreen1.1 mm75×25 mmLog in for availabilityCase of 1,440$520.25Supplier Number: 0314-7121-08Specifications:
More
Specifications
Cat. No.76204-212 - Slides, super colored glass, beveled edges, coloredsingle frosted one end, 45° clipped corners, printablePurple1.1 mm75×25 mmLog in for availabilityCase of 1,440$520.25Supplier Number: 0314-7121-09Specifications:
More
Specifications
Cat. No.76204-214 - Slides, super colored glass, beveled edges, colored single frosted one end, 45° clipped corners, printablePink1.1 mm75×25 mmLog in for availabilityCase of 1,440$520.25Supplier Number: 0314-7121-12Specifications:
More
Specifications
Cat. No.76204-216 - Slides, super colored glass, beveled edges, colored single frosted one end, 45° clipped corners, printableWhite1.1 mm75×25 mmLog in for availabilityCase of 1,440$520.25Supplier Number: 0314-7121-16Specifications:
More
Specifications
Cat. No.76204-218 - Slides, super colored glass, beveled edges, colored single frosted one end, 45° clipped corners, printableYellow1.1 mm75×25 mmLog in for availabilityCase of 1,440$520.25Supplier Number: 0314-7121-17Specifications:
More
Specifications
Cat. No.76204-220 - Slides, super grade, ground edges, colored single frosted one end, 45° clipped corners, printableWhite1.1 mm75×25 mmLog in for availabilityCase of 1,440$391.21Supplier Number: 0302-7121-16Specifications:
More
Specifications
Cat. No.76204-248 - Slides, super grade, ground edges, colored single frosted one end, 45° clipped corners, printableYellow1.1 mm75×25 mmLog in for availabilityCase of 1,440$391.21Supplier Number: 0302-7121-17Specifications:
More
Specifications
Cat. No.76204-250 - Slides, super grade, ground edges, plain, 90° cornersPlain1.1 mm75×25 mmDiscontinued-Supplier Number: 0303-0022Specifications:
More
Specifications
Cat. No.76204-252 - Slides, super grade, ground edges, single frosted one end, 90° cornersPlain1.1 mm75×25 mmLog in for availabilityCase of 1,440$335.35Supplier Number: 0303-2122Specifications:
More
Specifications
Cat. No.76204-254