Nylon Conductive Tapes
Nylon Conductive Tapes
Supplier:   BROWN DOG GADGETS
Nylon Conductive Tapes
Supplier:  BROWN DOG GADGETS
Certificates
Nylon Conductive Tapes
Nylon Conductive Tapes
2 Options Available Below
Product Details & Documents
Introducing: Maker Tape!
  • Blend of nylon and steel
  • Low resistance per foot
  • Strong and flexible, tear resistant
  • Overlapping layers make conducting electrical connection
Our favorite nylon-steel blend is now thinner and z-axis conductive. Just overlap two pieces of Maker Tape to create a solid electrical connection. This tape is much stronger and easier to work with than commonly used copper foil tapes. The ⅛" width tape is perfect for Crazy Circuits™ projects on LEGO™ bricks or compatible systems. The ¼" width is perfect for sticking down LEDs in Paper Circuits projects.
Ordering information: The ⅛" × 5 m tape is about 16' long. ¼" × 20 m tape is about 65' long.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Conductive maker tape, ⅛" × 5 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 8,70 $
    8,70 $
    Each
       8,70 $
    Supplier Number: P2029-05M
  • Conductive Maker Tape™, ¼" × 20 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 16,45 $
    16,45 $
    Each
       16,45 $
    Supplier Number: P2030-20M