Paper Circuits Kit
Paper Circuits Kit
Supplier:   BROWN DOG GADGETS
Paper Circuits Kit
Supplier:  BROWN DOG GADGETS
Certificates
Paper Circuits Kit
2 Options Available Below
Product Details & Documents
Perfect for putting the 'A' in STEAM, why mess with wires when you can use conductive tape?
  • Includes jumbo LEDs and conductive maker tape to make your project shine!
  • Perfect for makers of all ages
  • No soldering or special tools needed
Paper circuits are made with LEDs, coin cell batteries, conductive maker tape, and everyday craft supplies. Use templates available online or create your own designs to light up a craft, poster or tri-fold presentation board.
Paper circuits are economical, engaging and a great tool for hands-on learning of simple circuitry. The standard kit contains materials for at least 10 projects. The classroom kit contains materials for at least 25 projects.
Ordering information: Standard kit includes (30) jumbo LEDs; (12) binder clips; (10) CR2032 coin cell batteries; (1) roll of ¹/₄"×5 m maker tape; access to online resources.
Classroom kit includes (120) jumbo LEDs; (40) binder clips; (40) CR2032 coin cell batteries; (2) rolls of ¹/₄"×20 m maker tape; access to online resources.
Product Family Options
Informations sur le produit
  • Désignation
    No. of Students
    Availability
    Price
  • Standard kit
    10
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $44.10
    $44.10
    Each
       $44.10
    Supplier Number: K1006-STA
  • Classroom set
    25
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $176.55
    $176.55
    Each
       $176.55
    Supplier Number: K1006-CLS
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Conductive Maker Tape™, ¼" × 20 m
    Conductive Maker Tape™, ¼" × 20 m
    470234-666
    Log in pour la disponibilité
    Each - $16.45
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Conductive maker tape, ⅛" × 5 m
    Conductive maker tape, ⅛" × 5 m
    470234-664
    Log in pour la disponibilité
    Each - $8.70