BristleBot Kit
BristleBot Kit
Supplier:   BROWN DOG GADGETS
BristleBot Kit
Supplier:  BROWN DOG GADGETS
Certificates
BristleBot Kit
3 Options Available Below
Product Details & Documents
Teach STEAM with ease!
  • Build time 5 to15 minutes
  • Age range 9+
  • Tools needed: none
Bristlebots are small robots made by combining a toothbrush head and vibrating motor. By changing the position of the pipe cleaner arms, Bristlebots will travel in circles, straight lines, or scoot around randomly. What can you make your Bristlebot do? Bristlebots are great for teaching engineering, motors, circuits, and principles of balance.
Ordering information: Bristlebots do not require tools to build. The toothbrush head comes pre-cut, and motor wires have exposed ends to connect to the battery.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Bristlebots, Pack of 4
    Log in for availability
    Log in for availability
    Pack of 4
    Pack of 4 - $44.99
    $44.99
    Pack of 4
       $44.99
    Supplier Number: K1003-4PK
  • Bristlebots, Pack of 10
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $86.99
    $86.99
    Pack of 10
       $86.99
    Supplier Number: K1003-10PK
  • Bristlebots, Pack of 25
    Log in for availability
    Log in for availability
    Pack of 25
    Pack of 25 - $176.55
    $176.55
    Pack of 25
       $176.55
    Supplier Number: K1003-25PK