Sf9 Cells
Sf9 Cells
Sf9 Cells
Certificates
4 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Expression Systems offers insect cell lines adapted to serum-free suspension culture in either ESF 921 or ESF AF.
  • Recombinant protein expression from a variety of expression platforms
  • Good growth in adherent or suspension culture
  • Small, regular size that generates even monolayers and plaques
  • Quality and performance testing
The Sf9 insect cell line is a clonal isolate derived from the parental Spodoptera frugiperda cell line IPLB-Sf-21-AE. Sf9 cells are adapted for suspension culture in ESF 921 or ESF AF and are available as a frozen vial or suspension culture. Sf9 cells are commonly used to produce recombinant baculoviral stocks and to produce recombinant proteins.
The availability of pre-adapted cells saves significant time and expense associated with the adaptation of cultures.
Culture suspension shipments immediately, liquid nitrogen (vapor phase) for frozen vials.
Delivery information: Priority overnight.
Product Family Options
Product Information
  • Désignation
    Format de cellule
    Availability
    Price
  • Sf9 cells, frozen in ESF 921, 50 M cells/vial
    Cryopreserved
    Log in for availability
    Log in for availability
    Each
    Each - 857,66 $
    857,66 $
    Each
       857,66 $
    Minimum order quantity is 1
    Supplier Number: 94-001F
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Sf9 cells, frozen in ESF AF, 50 M cells/vial
    Cryopreserved
    Log in for availability
    Log in for availability
    Each
    Each - 866,87 $
    866,87 $
    Each
       866,87 $
    Minimum order quantity is 1
    Supplier Number: 94-006F
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Sf9 cells, suspension in ESF 921, approx. 1 M cells/ml (50/125 ml flask)
    Proliferative
    Log in for availability
    Log in for availability
    Each
    Each - 386,83 $
    386,83 $
    Each
       386,83 $
    Minimum order quantity is 1
    Supplier Number: 94-001S
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Sf9 cells, suspension in ESF AF, approx. 1 M cells/ml (50/125 ml flask)
    Proliferative
    Log in for availability
    Log in for availability
    Each
    Each - 386,83 $
    386,83 $
    Each
       386,83 $
    Minimum order quantity is 1
    Supplier Number: 94-006S
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.