Order Entry
Canada
ContactUsLinkComponent
Accessories for Fastprep Sample Preparation Instrument TallPrep™, MP Biomedicals
  CA76045-842
Accessories for Fastprep Sample Preparation Instrument TallPrep™, MP Biomedicals
  CA76045-842
 :  116002540
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

 

  • Description:
    TallPrep™ Adapter, 24×4.5 ml
  • Cat. no.:
    CA76045-842

 

 

TallPrep™ 24×4.5 ml Adapter is ideally suited for high-throughput applications, in which up to 24 samples can be homogenized simultaneously.

  • Ergonomic design
  • Easy loading of 4.5 ml tubes
  • Stands stable on the bench top
  • Commonly used as a tube rack

With the FastPrep-24, or FastPrep-24 5G Instrument, TallPrep adapter and optimized Lysing Matrices, consistent reproducible results are obtained.

Frozen samples in Lysing Matrix tubes loaded in the adapter and stored at –20 °C or –80 °C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.

Use with FastPrep-24, FastPrep-24 5G, or FastPrep-96 (with ConeFlex Legacy Adapter), MP Bio Lysing Matrix Tubes and/or FastPrep purification kits to lyse, grind or homogenize samples at ambient temperatures, for the extraction and purification of nucliec acids, proteins, metabolites and other small molecules and more.