Solid Caps, GL-Threads, Polypropylene, Chemglass
Supplier: Chemglass
Jump to:
À propos de cet article
These solid polypropylene caps can be autoclaved in steam to 140 °C.
9 Options Available Below
Product Details & Documents
These solid polypropylene caps can be autoclaved in steam to 140 °C.
Product Family Options
Informations sur le produit
- DésignationColorEpaisseurAvailabilityPrice
- Supplier Number: CLS-1480-10
- Supplier Number: CLS-1480-11
- Supplier Number: CLS-1480-12
- Supplier Number: CLS-1480-13SC
- Supplier Number: CLS-1480-14
- Supplier Number: CLS-1480-15
- Supplier Number: CLS-1480-16
- Supplier Number: CLS-1480-17
- Supplier Number: CLS-1480-18