YMC-Pack C4 (Butyl) HPLC Columns, YMC America
YMC-Pack C4 (butyl) columns are less hydrophobic than C8 or C18 packings, and generally utilize more aqueous eluents than either of these reversed-phase column types.
- Fully endcapped
- Ideally suited for biological separations
- High-coverage monomeric bonding on perfectly spherical silica supports
- Available in 120, 200 and 300 Å pore size packings
When compared to C8 and C18 columns (using the same eluents), C4 stationary phases show significantly shorter retention for non-polar compounds. Retention of polar compounds, however, is not significantly affected. Therefore, mixtures with a wide range of component polarity are best analyzed by YMC-Pack C4. This is due to the C4 bonded phase exhibiting shorter analysis time while still maintaining high resolution when compared to longer-chain bonded chemistries.
- Catalog No:
- 97029-884
- 97030-124
- 97030-122
- 97029-812
- 89216-590
- 89216-588
- 89216-586
- 97030-112
- 97030-108
- 97029-866
- 97029-864
- 97030-120
- 97030-118
- 97029-876
- 97029-874
- 97030-116
- 97030-114
- 97029-872
- 97029-832
- 97029-830
- 97029-828
- 97029-840
- 97030-148
- 89216-620
- 97029-838
- 97030-146
- 89216-618
- 97029-836
- 89216-622
- 89216-610
- 89216-616
- 97029-822
- 89216-608
- 89216-606
- 97030-134
- 97030-132
- 97030-130
- 97029-820
- 97030-128
- 97029-886
- 97030-140
- 97030-138
- 97030-136
- 97029-854
- 97030-008
- 97030-006
- 97029-850
- 97029-848
- 97030-002
- 97029-772
- 97029-860
- 97030-014
- 97029-770
- 97029-858
- 97030-012
- 97029-856
- 97030-010
- 97029-846
- 97029-844
- 97029-842
- 97030-000
- 97030-150
- 97030-158
- 97029-782
- 97029-862
- 97030-102
- 89198-940
- 97030-100
- 97029-780
- 97029-778
- 97029-776
- 97030-092
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 - 7.5
- 2 – 7.5
- 2 – 7.5
- 2 - 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 - 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 – 7.5
- 2 - 7.5
- 2 - 7.5
- 2 - 7.5
- 2 – 7.5
- USP code:
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
Frequently Bought Together