Zerostat 3 Anti-Static Device, Bellex International
Catalogue # MSPP-Z108812
Fournisseur: BELLEX INTERNATIONAL MS
Spécifications
- Description:Zerostat 3 Anti Static Device
- Cat. no.:MSPP-Z108812
- Supplier No.:Z108812
Spécifications
À propos de cet article
The pistol-shaped molded plastic device with a trigger mechanism incorporates a unique piezo crystal device.
- No batteries or electrical power supply required
- Non-radioactive, convenient, hand-held and lightweight
- Constructed from durable hard plastic, a long lasting alternative to cleaning brushes
Prevents charged particles from flying during weighing or transfer of powders and keeps powders from sticking to containers. Effective in neutralizing electrostatic charges on virtually any surface and eliminates static electricity in the laboratory, plant, darkroom or home. Keeps film, glass, laboratory slides and plasticware dust free. Neutralizes static from photographic lenses, negatives and slides to release dust and lint.