Lauda® Microcool Circulation Chillers, MC Series, Brinkmann™
Fournisseur: Lauda-Brinkmann
Certificats
À propos de cet article
Microcool circulation chiller line with five compact models and cooling capacities from 0.25 to 1.2 kW.
- User interface with large LED display and membrane keyboard
- Autostart timer and auto-shutdown function
- Illuminated window for checking heat transfer liquid level
- Block pump with magnetic coupling of pump and electric motor
- Integrated overflow connection
- RS-232 interface and alarm contact standard
- Integrated filling funnel on top of the device
- Easily removable front grid
The compact MC 250 and MC 350 makes them ideal for being positioned on the benchtop
The compact MC 600, MC 1200 and MC 1200 W cooling capacity models are floor standing instruments to fit underneath the lab bench. They are equipped with a pressure gauge to display the pressure and casters which can be controlled and locked. Pump pressure can be adjusted via the integrated bypass.
The most powerful device is also available in a water-cooled version as the MC 1200 W.
The circulation chillers are equipped with a magnetic coupling pump. This supplies a pump pressure of 0.35 bar and a maximumpump flow of 16 L/min.
Frequently Bought Together