Ward’s® Adjustable Volume Micropipettes
Ward’s® Adjustable Volume Micropipettes
Ward’s® Adjustable Volume Micropipettes
Certificates
Ward’s® Adjustable Volume Micropipettes
Ward’s® Adjustable Volume Micropipettes
9 Options Available Below
Product Details & Documents
The Wards® Micropipette line is fully autoclavable, helping to prevent cross contamination. The ergonomic design is ambidextrous, providing all users with an excellent operating experience.
  • Fully autoclavable
  • Slim, lightweight ergonomic design
  • Compatible with most universal tip brands
  • Tool included for quick and easy recalibration
  • Purchase individually, or as one of our available sets
Personal Pipette Set comes with one of each pipette shown, plus a storage carousel.
Classroom Pipette Set comes with eight of each pipette shown, plus eight storage carousels.
Product Family Options
Micropipettes
Product Information
  • Désignation
    Volume
    Increments
    Availability
    Price
  • 10 μL Micropipette
    0.5 - 10 µl
    0.1 µl
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $289.75
    $289.75
    Log in for availability
    Box of 50
    Box of 50 - $5,856.70
    $5,856.70
    Each
       $289.75
    Box of 50
       $5,856.70
    Supplier Number: 7010301004
  • 20 μL Micropipette
    2 - 20 µl
    0.5 µl
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $263.40
    $263.40
    Log in for availability
    Box of 50
    Box of 50 - $5,856.70
    $5,856.70
    Each
       $263.40
    Box of 50
       $5,856.70
    Supplier Number: 7010301005
  • 100 μL Micropipette
    10 - 100 µl
    1.0 µl
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $289.75
    $289.75
    Log in for availability
    Box of 50
    Box of 50 - $5,856.70
    $5,856.70
    Each
       $289.75
    Box of 50
       $5,856.70
    Supplier Number: 7010301008
  • 200 μL Micropipette
    20 - 200 µl
    1.0 µl
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $289.75
    $289.75
    Log in for availability
    Box of 50
    Box of 50 - $5,856.70
    $5,856.70
    Each
       $289.75
    Box of 50
       $5,856.70
    Supplier Number: 7010301009
  • 1000 μL Micropipette
    100 - 1000 µl
    5.0 µl
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $289.75
    $289.75
    Log in for availability
    Box of 50
    Box of 50 - $5,856.70
    $5,856.70
    Each
       $289.75
    Box of 50
       $5,856.70
    Supplier Number: 7010301014
  • 5000 uL Micropipette
    1000 - 5000 µl
    50.0 µl
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $263.40
    $263.40
    Log in for availability
    Box of 50
    Box of 50 - $5,856.70
    $5,856.70
    Each
       $263.40
    Box of 50
       $5,856.70
    Supplier Number: 7010301017
  • Personal Pipette Set
    Log in for availability
    Log in for availability
    Each
    Each - $1,066.50
    $1,066.50
    Each
       $1,066.50
    Supplier Number: -
  • Classroom Pipette Set
    Log in for availability
    Log in for availability
    Each
    Each - $6,975.00
    $6,975.00
    Each
       $6,975.00
    Supplier Number: -
Micropipette Holder
Product Information
  • Désignation
    Volume
    Increments
    Availability
    Price
  • Micropipette Carousel (holds 6)
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $121.35
    $121.35
    Log in for availability
    Box of 10
    Box of 10 - $1,077.25
    $1,077.25
    Each
       $121.35
    Box of 10
       $1,077.25
    Supplier Number: 7010000084
Micropipette Tips
Product Information
  • Catalog #
    Packed
    Volume
    Tip Color
    Availability
    Price
  • 100 Tips/Bag
    1000 - 5000 µl
    Log in for availability
    Log in for availability
    Bag of 100
    Bag of 100 - $83.75
    $83.75
    Bag of 100
       $83.75