Caps, BASIK™, Crimp for ND8, MicroSolv
Supplier: MicroSolv Technology Corp
Jump to:
À propos de cet article
These caps are suitable for 8 mm Crimp top Vials.
- Center Hole: 4 mm
2 Options Available Below
Product Details & Documents
These caps are suitable for 8 mm Crimp top Vials.
- Center Hole: 4 mm
Product Family Options
Informations sur le produit
- DésignationÉpaisseurCap TypeAvailabilityPrice
- Crimp Caps, ND8, Natural Rubber/ PTFE—PEDiscontinued-Supplier Number: 9508C-10C-C
- Crimp Caps, ND8, 60 Shore A, Natural Rubber/TEF Septa1 mmAluminum capLog in pour la disponibilitéPack of 100$37.38Supplier Number: 9508C-40T-S
- Crimp Caps, ND8, Silicone Rubber/ Red PTFE—PEDiscontinued-Supplier Number: 9508C-10C-D
- Crimp Caps, ND8, 45 Shore A, Sil/PTFE with preslit1.3 mmAluminum capLog in pour la disponibilitéPack of 100$140.17Supplier Number: 9508C-30T-S