Pediatric Micro Sample Cups, Globe Scientific
Pediatric Micro Sample Cups, Globe Scientific
Pediatric Micro Sample Cups, Polystyrene
Catalog # 75982-638
Supplier:   Globe Scientific
Supplier #:  110915
Pediatric Micro Sample Cups, Globe Scientific
Catalog # 75982-638
Supplier:  Globe Scientific
Pediatric Micro Sample Cups, Globe Scientific
Pediatric Micro Sample Cups, Globe Scientific
Pediatric Micro Sample Cups, Polystyrene
Price
$174.77
New Customer Promotional Price
Log in for availability
Bag of 1,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This polystyrene (PS) micro sample cup is designed for use on automated instrumentation including the Hitachi®, Elecsys®, Cobas Integra®, and Core® analyzers.
Specifications
  • Désignation:
    Pediatric Micro Sample Cups
  • Matière:
    Polystyrene