ContactUsLinkComponent
SP Bel-Art Tall Form Graduated Pitchers, Clear TPX, Bel-Art Products, a part of SP
SP Bel-Art Tall Form Graduated Pitchers, Clear TPX, Bel-Art Products, a part of SP
Catalog # 13197-210
Supplier #: F28982-0000
undefined
SP Bel-Art Tall Form Graduated Pitchers, Clear TPX, Bel-Art Products, a part of SP
Catalog # 13197-210
Supplier Number:  F28982-0000
Supplier #: F28982-0000
Log in for availability
Select an Option
76 mm (3") Bottom; 95 mm (3<sup>3</sup>/<sub>4</sub>") Top
Select an Option
10 ml
Select an Option
500 ml
Select an Option
133 mm (5¼")
Select an Option
13197-210
Select an Option
F289820000
About this item
Gracefully proportioned, these pitchers are ideal for general laboratory use and offer well designed spouts and large handles helpful for use with wet or dry ingredients.
  • Crystal clear Polymethylpentene (TPX®)
  • Chemical resistant and shatterproof
  • Heat resistant to 175 °C (347 °F)
  • Steam autoclavable
Metric graduations molded on outside wall.
Frequently Bought Together