Caps for Sample Cups, Globe Scientific
About this item
Produced from polyethylene (PE), these caps are designed specifically for use on Globe's popular line of multi-purpose sample cups
Suitable for use with the Globe Scientific Multi-Purpose Sampling Cups series.
Caps are available with either a solid or cross-cut pierceable top, and they snap securely in place on each of the sample cups.
2 Options Available Below
Product Details & Documents
Produced from polyethylene (PE), these caps are designed specifically for use on Globe's popular line of multi-purpose sample cups
Suitable for use with the Globe Scientific Multi-Purpose Sampling Cups series.
Caps are available with either a solid or cross-cut pierceable top, and they snap securely in place on each of the sample cups.
Product Family Options
Product Information