Microscope Slide Mailer for 5 Slides with Side Opening, Globe Scientific
75845-146
- Description:Polypropylene Slide Mailer with Side Opening
- Cat. no.:75845-146
- Supplier No.:513065
The side opening lid provides easy access to the entire length of the slide (not just the end)
Slide mailers are produced from polypropylene and feature inner slots which keep slides separated and secure during transport.