Packaging Buckets, Burkle
Packaging Buckets, Burkle
Supplier:   BURKLE INC
Packaging Buckets, Burkle
Supplier:  BURKLE INC
Certificates
Packaging Buckets, Burkle
Packaging Buckets, Burkle
5 Options Available Below
Product Details & Documents
Packaging buckets for transport, packaging, and storing of various materials
  • Polypropylene, white finish
  • Air-tight closure
  • Supplied with lid
  • Autoclavable
  • Also available with advertising print (in large quantities on request)
Suitable for the food or building material industries and non-aggressive chemicals in environmental protection, hospitals, and the household. Buckets are equipped with a strong handle and a quality closure for product safety.
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Diameter
    Hauteur
    Availability
    Price
  • PP packaging bucket
    1 L (0.26 gal.)
    132 mm (5.20")
    131 mm (5.2") with cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5.60
    $5.60
    Each
       $5.60
    Supplier Number: 2327-0001
  • PP packaging bucket
    2 L (0.5 gal.)
    175 mm (6.89")
    132 mm (5.2") with cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8.90
    $8.90
    Each
       $8.90
    Supplier Number: 2327-0020
  • PP packaging bucket
    3 L (0.79 gal.)
    225 mm (8.86")
    123 mm (4.8") with cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $11.30
    $11.30
    Each
       $11.30
    Supplier Number: 2327-0030
  • PP packaging bucket
    5 L (1.3 gal.)
    225 mm (8.86")
    194 mm (7.6") with cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $14.40
    $14.40
    Each
       $14.40
    Supplier Number: 2327-0050
  • PP packaging bucket
    10 L (2.6 gal.)
    268 mm (10.55")
    263 mm (10.35") with cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $18.64
    $18.64
    Each
       $18.64
    Supplier Number: 2327-0010