Digi-Sense® Benchtop Variable Voltage Output Controller, Cole-Parmer
Digi-Sense® Variable-Voltage Output Controller, 5 − 100%, 120 V, 10 A
Catalog # 76183-394
Supplier: Antyila Scientific
Fournisseur #: 36225-59
Digi-Sense® Benchtop Variable Voltage Output Controller, Cole-Parmer
Catalog # 76183-394
Supplier: Antyila Scientific
Jump to:
Product Details & Documents
Designed to regulate the input voltage through the selector knob.
- Works with resistive loads, providing variable voltage
- Simple to use; no programming or feedback required
- With reference knob (0 to 100%)
Output 1: Proportional Voltage
Ordering information: Includes 6 ft. attached power cord.
Spécifications
- Désignation:Digi-Sense® Variable-Voltage Output Controller, 5 − 100%, 120 V, 10 A