Wallcur® PRACTI-Insulin
Wallcur® PRACTI-Insulin
Wallcur® PRACTI-Insulin
Wallcur® PRACTI-Insulin
Practice With A Wide Variety Of Insulin Products
  • Contains No Medication
  • Multiple Options Available
Teaching insulin injections starts with the fundamentals of carefully reading the insulin vial labels for dosage information as many insulin labels look alike. With PRACTI- Insulin you can teach your students how to calculate dosages, choose the applicable syringe for the dosage, draw up the correct amount of air, and then rotate, mix and withdraw before the medication precipitates out. Wallcur’s PRACTI- Insulin Vials contain distilled water and an additive that closely simulates the real thing and must be mixed properly, and administered promptly.
Teach your students to:
Identify and understand two of the most commonly ordered insulin medications
Read and interpret differing insulin labels that often look-a-like
Calculate insulin dosages
Mix, combine, and promptly withdraw short and long acting insulin
Identify injection site landmarks, and aseptic precautions when administering insulin
 : These products require a release form for purchase. If your account does not have one on file we can not ship until we receive your release documentation.
 : For Training Purposes only, not for Human or Animal use
Product Family Options
  • Désignation
    Includes
  • PRACTI-Novlog (Aspart) Insulin Pack
    Distilled Water
    Box of 40
    Box of 40 - 134,49 $
    134,49 $
    Box of 40
       134,49 $
     : 445NL
  • PRACTI-NPH Insulin Vial
    Barium Sulfate And Distilled Water
    Box of 40
    Box of 40 - 136,49 $
    136,49 $
    Box of 40
       136,49 $
     : 9405NPH
  • Practi-Insulin detemir
    Unsterile fructose powder
    Box of 40
    Box of 40 - 146,94 $
    146,94 $
    Box of 40
       146,94 $
     : 306DT
  • PRACTI-Regular Insulin Vial
    Distilled Water
    Box of 40
    Box of 40 - 136,49 $
    136,49 $
    Box of 40
       136,49 $
     : 9405REG
  • PRACTI-70/30 Insulin Vial
    Barium Sulfate And Water
    Box of 40
    Box of 40 - 134,49 $
    134,49 $
    Box of 40
       134,49 $
     : 414VP
  • PRACTI-Insulin lispro (Humalog)
    Distilled water
    Box of 40
    Box of 40 - 134,49 $
    134,49 $
    Box of 40
       134,49 $
     : 452HMG
  • PRACTI-Insulin Starter Pack Vials And 12 Labels
    Barium Sulfate And Water
    Kit of 1
    Kit of 1 - 125,39 $
    125,39 $
    Kit of 1
       125,39 $
     : SP-419
  • PRACTI-Insulin Variety Pack 10 NPH, 10 Reg, 1070/30, 10 Lantuz
    Barium Sulfate And Water
    Kit of 1
    Kit of 1 - 134,49 $
    134,49 $
    Kit of 1
       134,49 $
     : 408VP
  • PRACTI-Insulin Training Pack 20 NPH And 20 Reg
    Barium Sulfate And Distilled Water
    Box of 40
    Box of 40 - 134,49 $
    134,49 $
    Box of 40
       134,49 $
     : 405TP
  • PRACTI-HUMLN Insulin Pack 20 R And 20 N
    Barium Sulfate And Water
    Box of 40
    Box of 40 - 134,49 $
    134,49 $
    Box of 40
       134,49 $
     : 409HM
  • PRACTI-Insulin 70/30™ and glargine pack
    Barium sulfate and water
    Box of 40
    Box of 40 - 134,49 $
    134,49 $
    Box of 40
       134,49 $
     : 47030
  • PRACTI-Insulin glargine (Lantus)
    Distilled water
    Box of 40
    Box of 40 - 154,49 $
    154,49 $
    Box of 40
       154,49 $
     : 410LT