Wallcur® PRACTI-Nitroglycerin Spray
Wallcur® PRACTI-Nitroglycerin Spray
PRACTI-Nitroglycerin Spray, Cfc-Free Aerosol Propellant
Catalog # 470233-032
Supplier:   WALLCUR, LLC.
Supplier #:  720NS
Wallcur® PRACTI-Nitroglycerin Spray
Catalog # 470233-032
Supplier:  WALLCUR, LLC.
Wallcur® PRACTI-Nitroglycerin Spray
Wallcur® PRACTI-Nitroglycerin Spray
PRACTI-Nitroglycerin Spray, Cfc-Free Aerosol Propellant
Price
92,39 $
Log in pour la disponibilité
Pack of 5
Product Details & Documents
For Clinical Training
  • Contains No Medication
  • Over 200 Sprays Per Container
PRACTI-Nitroglycerin Spray™ is a CFC free pump that simulates a sublingual aerosol spray. Labeled identical to its true drug counterpart, nitroglycerin, which is used to treat episodes of angina in people who have coronary artery disease. Each spray pump looks, acts, and feels like the real thing! Contains no medication and provides 200 sprays per pump. PRACTI-Nitroglycerin Spray™ is a helpful and effective addition to your current patient care scenarios.
Pre-clinical learning includes:
Reading medication labels
Calculating the dosage
Priming the pump for use
Delivery information: These products require a release form for purchase. If your account does not have one on file we can not ship until we receive your release documentation.
Caution: For Training Purposes only, not for Human or Animal use
Spécifications
  • Désignation:
    PRACTI-Nitroglycerin Spray
  • Includes:
    Cfc-Free Aerosol Propellant