Wallcur® PRACTI-Inhalers
About this item
Train Your Students To Train Patients!
- Contains No Medication
- Available In Either A Generic, Or Labeled Trainer
Wallcur has the only inhaler available with just an aerosol propellant developed specifically for clinical training. Each PRACTI-Inhaler™ is equipped to simulate medication delivery in your practice skills lab. Excellent for Practicing verbal instruction and demonstrating hand/breath coordination. Also available in a proAyer and Ipratropium version allowing trainees to learn adjustment and dosage control.
Delivery information: These products require a release form for purchase. If your account does not have one on file we can not ship until we receive your release documentation.
Caution: For Training Purposes only, not for Human or Animal use
3 Options Available Below
Product Details & Documents
Train Your Students To Train Patients!
- Contains No Medication
- Available In Either A Generic, Or Labeled Trainer
Wallcur has the only inhaler available with just an aerosol propellant developed specifically for clinical training. Each PRACTI-Inhaler™ is equipped to simulate medication delivery in your practice skills lab. Excellent for Practicing verbal instruction and demonstrating hand/breath coordination. Also available in a proAyer and Ipratropium version allowing trainees to learn adjustment and dosage control.
Delivery information: These products require a release form for purchase. If your account does not have one on file we can not ship until we receive your release documentation.
Caution: For Training Purposes only, not for Human or Animal use
Product Family Options
Product Information
- DésignationIncludesAvailabilityPrice
- Supplier Number: 619PISpecifications:
- PRACTI-Ipratropium 18 mcg/Puff InhalerCvc-Free Aerosol PropellantLog in for availabilityPack of 5$94.39Supplier Number: 667TRSpecifications:
- Supplier Number: 669PASpecifications: