Wallcur® PRACTI-Naloxone
Wallcur® PRACTI-Naloxone
Supplier:   WALLCUR, LLC.
Wallcur® PRACTI-Naloxone
Supplier:  WALLCUR, LLC.
Certificates
Wallcur® PRACTI-Naloxone
Wallcur® PRACTI-Naloxone
2 Options Available Below
Product Details & Documents
Increase Your Students' Naloxone Administration Knowledge!
  • Contains No Medication
  • Available In Either A Vial, Or Prefilled Syringe
  • Contents Are Safe For Use With Most Simulators
PRACTI-Naloxone HCI™ for clinical training. Provide your students with the most realistic and authentic practice necessary to master the skill of naloxone hydrochloride drug calculation and administration. In addition to being one of the most difficult-to-handle vial sizes, your students are now able to practice a variety of clinical scenarios related to opioid‐induced respiratory depression and overdose. Available in simulated vials, and prefilled syringes with nasal atomizer. A prefilled Code syringe is available with the PRACTI-Code Meds.
PRACTI-Naloxone HCI™ 0.4 mg/mL comes in a clear 1 mL vial with distilled water, allowing for safe practice with manikins and other injection simulators. PRACTI-Naloxone HCI™ is the perfect addition to your EMS, EMT, or Paramedic program.
PRACTI-Intranasal Naloxone™ Training Pack contains a (1 mg/mL) 2 mg per 2 mL pre-filled luer lock syringe and nasal atomizer device for clinical training only. Wallcur's PRACTI-Naloxone simulates the opioid antagonist drug, Naloxone for emergency opioid overdose. Teach a variety of skills to your students of nursing and EMT programs.
Delivery information: These products require a release form for purchase. If your account does not have one on file we can not ship until we receive your release documentation.
Caution: For Training Purposes only, not for Human or Animal use
Product Family Options
Product Information
  • Désignation
    Includes
    Availability
    Price
  • PRACTI-Naloxone HCl 1 mL Vial
    Distilled Water
    Log in for availability
    Log in for availability
    Box of 30
    Box of 30 - $121.69
    $121.69
    Box of 30
       $121.69
    Supplier Number: 438NX
    Specifications:
  • PRACTI-Intranasal Naloxone Training Pack
    Distilled Water
    Log in for availability
    Log in for availability
    Each
    Each - $34.19
    $34.19
    Each
       $34.19
    Supplier Number: 1517NA
    Specifications:
Recommendations