Choice® Edge Cleanroom-Laundered, Bordered, Sealed-Edge 100% Polyester Knit Wiper, Berkshire
Catalog # 76193-156
Supplier: Berkshire
Specifications
- Description:Ultrasonically Sealed
- Material:Knitted Polyester
- Cat. no.:76193-156
- Packaging:Stacked
- W×L:22.9×22.9 cm (9×9")
Specifications
About this item
Smooth knit construction and consistent border with sealed edges for scratch-free wiping.
- No-run interlock construction
- Sealed edges for increased durability
Wipers are made from 100% polyester knit. Four-side-bordered, with sealed edges to increase durability and minimize fiber and particle release.
Certifications: Laundered in an ISO Class 4 (Class 10) Cleanroom.