Somso® Catheterization Demonstration Models | Avantor
Somso® Catheterization Demonstration Models
Supplier:   MARCUS SOMMER
Somso® Catheterization Demonstration Models
Supplier:  MARCUS SOMMER
Certificates
Somso® Catheterization Demonstration Models
Somso® Catheterization Demonstration Models
2 Options Available Below
Product Details & Documents
Show your students what's happening.
  • Great for basic practice
  • Opens to show catheter in place
  • Male and Female versions available
Ideal for demonstrating disposable and balloon catheters as well as suprapubic aspiration. Can be dismantled into two parts to show the catheter in position. In natural size, made of special plastic. On a stand with green base.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Catheterization-Model, Male
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $462.00
    $462.00
    Each
       $462.00
    Supplier Number: CLA7/10
  • Catheterization-Model, Female
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $462.00
    $462.00
    Each
       $462.00
    Supplier Number: CLA7/14