Somso® OGI Phantom | Avantor
Somso® OGI Phantom
Supplier:   MARCUS SOMMER
Somso® OGI Phantom
Supplier:  MARCUS SOMMER
Certificates
Somso® OGI Phantom
Somso® OGI Phantom
2 Options Available Below
Product Details & Documents
Endoscopic practice in the Upper GI tract.
  • Multiple procedures can be practiced
  • Replicable stomach
Esophagoscopy, gastroscopy, bulboscopy as well as endoscopic-retrograde cholangiography-pancreaticography (ERCP) according to Prof. Dr. M. Classen, and H. Ruppin M.D. For training of endoscopy of the upper gastrointestinal tract, for catheterization of the papilla vateri by retrograde instillation of contrast medium into the pancreatic duct system.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • OGI Phantom
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,070.00
    $6,070.00
    Each
       $6,070.00
    Supplier Number: CLA4
  • Stomach And Duodenum
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,270.00
    $1,270.00
    Each
       $1,270.00
    Supplier Number: CLA4/1