PYREX® Volumetric Flask with Glass [ST] Stopper, Class A, Corning
Supplier: Corning
Certificates
About this item
These PYREX® 5 ml volumetric flasks provide improved lab convenience, broad selection, and precise volume measurement.
- Type I, class A, low expansion borosilicate glass
- Class A accuracy
- Easy to read graduation
These class A volumetric flasks have been manufactured to class A tolerances as established by ASTM E- 694 for volumetric ware, ASTM E-542 for calibration of volumetric ware, and ASTM E-288 for volumetric flasks.
The strength of these flasks has been increased appreciably through machine-blown bodies, to which are sealed heavy-tubing necks tooled for No.8 standard taper glass stoppers. The graduation line is sharp and permanent and large white block letters make the inscription easy to read.
Certifications: Meets ASTM standards E-438, E-694, E-542, and E-288.
Specifications
- Catalog No:
- 89091-402
- 89090-752
- 89090-912
- 89091-480
- 89091-218
- 89091-062
- 89091-102
- 26307-462
- 89090-606
- 89090-714
- 89090-848
- 89090-680
- 89090-818
- 89090-770
- 29619-915
- 29619-926
- 89091-300
- 89091-094
- 26307-466
- 29619-970
- 26307-464
- 29619-948
- 26307-468
- 29619-959
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey