ContactUsLinkComponent
SensoLyte® ADHP Peroxidase Assay Kit (Fluorimetric), AnaSpec Inc.
Catalogue # 103010-128
Fournisseur:  Anaspec Inc
Numéro CAS:  
SensoLyte® ADHP Peroxidase Assay Kit (Fluorimetric), AnaSpec Inc.
Catalogue # 103010-128
Fournisseur:  Anaspec Inc
Supplier Number:  AS-71111
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    500 Assays
  • Cat. no.:
    103010-128
  • Storage Temperature:
    – 20 °C
  • Includes:
    The assays can be performed in a convenient 96-well or 384-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier No.:
    AS-71111
Spécifications
À propos de cet article
HRP conjugates are extensively used as secondary detection reagents in ELISAs, immuno-histochemical techniques and Northern, Southern and Western blot analyses.
SensoLyte® ADHP Peroxidase Assay Kit uses highly purified ADHP to quantify peroxidase activity in solutions, in cell extracts and in live cells and on solid surfaces (such as PVDF membranes). The kit can be used for characterizing kinetics of enzyme reaction and high throughput screening of oxidase inhibitors. The kit inlcludes ADHP peroxidase substrate (Ex/Em=573/590 nm upon oxidation) and an optimized ‘mix and read’ assay protocol that is compatible with HTS liquid handling instruments, among all essential assay components.