ContactUsLinkComponent
SensoLyte® 520 HDAC Activity Assay Kit (Fluorimetric), AnaSpec
SensoLyte® 520 HDAC Activity Assay Kit (Fluorimetric), AnaSpec
Catalog # 103008-960
Supplier:  Anaspec Inc
CAS Number:  
SensoLyte® 520 HDAC Activity Assay Kit (Fluorimetric), AnaSpec
Catalog # 103008-960
Supplier:  Anaspec Inc
Supplier Number:  AS-72084
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    100 Assays
  • Cat. no.:
    103008-960
  • Storage Temperature:
    – 78 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier No.:
    AS-72084
Specifications
About this item
Histone deacetylase (HDAC) enzymes modulate gene expression through the deacetylation of lysine residues on histone proteins and act as transcriptional repressors of genes
The SensoLyte® 520 HDAC Activity Assay Kit provides a convenient, two-step homogeneous procedure for measuring HDAC activity using a fluorogenic substrate. In the first step, an acetylated substrate is incubated with HDAC containing samples. Deacetylation of substrate sensitizes it, so that, in the second step mixing with the HDAC developer generates a fluorophore, which can then be detected at Ex/Em= 490 nm/520 nm. The substrate included in the kit is cell-permeable, and the assay can be used to measure HDAC activity directly in cell culture in a 96-well plate without a time-consuming cell extraction step. This kit also can be used for high throughput screening of HDAC inhibitors using extracts or purified enzymes. The long wavelength fluorescence of HDAC 520 substrate is less interfered by the autofluorescence of cell components and test compounds.
Based on their role in cell cycling, apoptosis and differentiation, HDACs have been chosen as therapeutic targets for the treatment of cancer and neurodegenerative diseases.