ContactUsLinkComponent
SensoLyte® GST Activity Assay Kit (Fluorimetric), AnaSpec Inc.
SensoLyte® GST Activity Assay Kit (Fluorimetric), AnaSpec Inc.
  103010-518
 :  
SensoLyte® GST Activity Assay Kit (Fluorimetric), AnaSpec Inc.
  103010-518
 :  AS-72157
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Size:
    100 Assays
  • Cat. no.:
    103010-518
  • Storage Temperature:
    – 78 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier No.:
    AS-72157
GST (Glutathione S-Transferase) enzymes, a family of isozymes plays a crucial role in body defense mechanisms against toxins and carcinogens
The SensoLyte® GST Activity Assay Kit utilizes an optimized method for detection of enzyme activity in biological samples. The kit provides substrate for GST enzyme, which upon reaction with reduced glutathione yields a fluorescent glutathione adduct. This produces bright blue fluorescence that can be monitored at excitation/emission =390 nm/480 nm.
GSTs carry out their functions mostly by mediating their conjugation with reduced glutathione on substrates such as pharmaceutical and environmental pollutants.