ContactUsLinkComponent
SensoLyte® 520 Aggrecanase-1 Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-186
Supplier:  Anaspec Inc
CAS Number:  
SensoLyte® 520 Aggrecanase-1 Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-186
Supplier:  Anaspec Inc
Supplier Number:  AS-72114
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    100 Assays
  • Cat. no.:
    103010-186
  • Storage Temperature:
    – 20 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier No.:
    AS-72114
Specifications
About this item
Aggrecanases belong to ADAMTS (A disintegrin and metalloprotease with thrombospondin motif) family of proteases
The SensoLyte® 520 Aggrecanase-1 Assay Kit uses 5-FAM as the fluorophore and TAMRA as the quencher in the FRET peptide substrate. This substrate is used for continuous measurement of the enzyme activity. In an intact FRET peptide, the fluorescence of 5-FAM is quenched by TAMRA. Upon the cleavage of the FRET peptide by ADAMTS-4, the fluorescence of 5-FAM is recovered, and can be continuously monitored at excitation/emission = 490 nm/520 nm. Increase in FAM fluorescence is correlated to aggrecanase-1 activity. 5-FAM/TAMRA based FRET substrate is less interfered by autofluorescence of test compounds due to its longer emission wavelength. The kit can be used to detect the activity of aggrecanase-1 or screen for aggrecanase inhibitors. The assays are performed in a convenient 96-well format.
Aggrecanases cleave aggrecan, the major structural component of cartilage. Aggrecanase-1 (ADAMTS-4) is a major aggrecanase in human osteoarthritic cartilage.