ContactUsLinkComponent
SensoLyte® Thioflavin T β-Amyloid (1-40) Aggregation Kit, AnaSpec
  103010-634
 :  Anaspec
 :  
SensoLyte® Thioflavin T β-Amyloid (1-40) Aggregation Kit, AnaSpec
  103010-634
 :  Anaspec
 :  AS-72213
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Size:
    100 Assays
  • Cat. no.:
    103010-634
  • Storage Temperature:
    +4 °C
  • Includes:
    Kit includes a 96-well plate, essential components and detailed protocol and references
  • Supplier No.:
    AS-72213
The SensoLyte® ThT β-Amyloid (1-40) Aggregation kit provides a convenient and standard method to measure Aβ40 aggregation using Thioflavin T dye
Alzheimer's disease (AD), the most common cause of dementia, is characterized by the presence of senile plaques and neurofibrillary tangles, surrounded by damaged neurons. Beta-Amyloid (Aβ) peptides, Aβ40 (1-40) and Aβ42 (1-42), were found to be a major component of the above plaques. Many studies suggest that these peptides can form toxic oligomers and fibrils under physiological conditions and rapidly aggregate. Since Aβ aggregation is evidently an essential event in the pathogenesis of AD, a reliable assay is important to study Aβ fibrillation kinetics and screen for Aβ aggregation inhibitors.
Aβ40 peptide is pretreated to ensure it is in a monomeric state. An optimized fibrillation buffer is included with the kit, and two known inhibitors are supplied as controls. The assay is based on the property of ThT dye in which fluorescence (Ex/Em=440/484 nm) is increased when bound to aggregated Aβ peptides.