Order Entry
Canada
ContactUsLinkComponent
SensoLyte® Generic MMP Assay Kit (Colorimetric), AnaSpec Inc.
Catalogue # 103010-412
Fournisseur:  Anaspec Inc
Numéro CAS:  
SensoLyte® Generic MMP Assay Kit (Colorimetric), AnaSpec Inc.
Catalogue # 103010-412
Fournisseur:  Anaspec Inc
Supplier Number:  AS-72095
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Spécifications

  • Size:
    100 Assays
  • Cat. no.:
    103010-412
  • Storage Temperature:
    –20 °C
  • Includes:
    The assays can be performed in a convenient 96-well or 384-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier No.:
    AS-72095

Spécifications

À propos de cet article

Matrix metalloproteinases (MMPs) belong to a family of secreted or membrane-associated proteins capable of digesting extracellular matrix components

The SensoLyte® Generic MMP Assay Kit can be used to detect the activity of a variety of MMPs, including MMP-1, 2, 3, 7, 8, 9, 12, 13, and 14 or for high throughput screening of MMPs’ inducers and inhibitors. The substrate is a thiopeptolide that is cleaved by the MMPs, releasing a sulfhydryl group, which can be detected with a color-developing thiol-reactive agent, 4,4’-dithiodipyridine or Ellman’s Reagent at 412 nm using a microplate reader. The assays are performed in a convenient 96-well microplate format.

The importance of MMPs in tumor development and invasion as well as other diseases is well known.