ContactUsLinkComponent
SensoLyte® 490 MMP-2 Assay Kit (Fluorimetric), AnaSpec Inc.
  103010-152
 :  
SensoLyte® 490 MMP-2 Assay Kit (Fluorimetric), AnaSpec Inc.
  103010-152
 :  AS-71129
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Size:
    500 Assays
  • Cat. no.:
    103010-152
  • Storage Temperature:
    –20 °C
  • Includes:
    The assays can be performed in a convenient 96-well or 384-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier No.:
    AS-71129
Matrix metalloproteinases (MMPs) belong to a family of secreted or membrane-associated proteins capable of digesting extracellular matrix components
The SensoLyte® 490 MMP-2 Assay Kit is a complete assay system designed to continuously analyze MMP activities or to screen MMP-2 inhibitors by using a fluorogenic MMP-2 substrate. The assays are performed in a convenient 96-well or 384-well microplate format. The kit provides speed, convenience and accuracy in analyzing MMP-2 activities and in screening MMP-2 inhibitors.
The importance of MMPs in tumor development and invasion is well known. MMP-2 (gelatinase A) is proposed as a potential therapeutic target.