To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Size:500 Assays
- Cat. no.:103010-152
- Storage Temperature:–20 °C
- Includes:The assays can be performed in a convenient 96-well or 384-well microplate format, kit includes essential components and detailed protocol and references
- Supplier No.:AS-71129
Matrix metalloproteinases (MMPs) belong to a family of secreted or membrane-associated proteins capable of digesting extracellular matrix components
The SensoLyte® 490 MMP-2 Assay Kit is a complete assay system designed to continuously analyze MMP activities or to screen MMP-2 inhibitors by using a fluorogenic MMP-2 substrate. The assays are performed in a convenient 96-well or 384-well microplate format. The kit provides speed, convenience and accuracy in analyzing MMP-2 activities and in screening MMP-2 inhibitors.
The importance of MMPs in tumor development and invasion is well known. MMP-2 (gelatinase A) is proposed as a potential therapeutic target.