Ward's® Meniscus Reader
Ward's® Meniscus Reader
New Product
Supplier:   VWR International
Ward's® Meniscus Reader
Supplier:  VWR International
New Product
Certificates
Ward's® Meniscus Reader
Ward's® Meniscus Reader
2 Options Available Below
Product Details & Documents
Easily see the endpoint of your titration.
  • Provides background that is white above and black beneath
  • Transparent front strap holds reader in place
  • Waterproof - flexible plastic card, not laminated paper
The Ward's® Meniscus reader makes it vastly easier to visualize the bottom of the meniscus in burets and small graduated cylinders. The contrasting black and white background allows students to easily see the meniscus in the buret - AND accurately read the volume. Slide the meniscus reader up or down to position the black demarcation just below the meniscus.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Lecteur de ménisque
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $17.30
    $17.30
    Each
       $17.30
  • Ward's® meniscus reader
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $11.46
    $11.46
    Each
       $11.46
    Spécifications:
    New Product