Freeze Substitution Kit, Electron Microscopy Sciences | Avantor
Freeze Substitution Kit, Electron Microscopy Sciences
Freeze Substitution Kit, Electron Microscopy Sciences
Certificates
Freeze Substitution Kit, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For dehydration and fixation of rapidly frozen cells.
  • Excellent freeze substitution results in as little as 90 minutes
  • Extend the time for larger samples to 3 hours by putting the lid on
Freeze Substitution is a process for low temperature dehydration and fixation of rapidly frozen cells that usually takes days to complete.
With this unique kit researchers are now able to achieve excellent freeze substitution results in as little as 90 minutes for cells of small volume such as bacteria and tissue culture cells. For those cells of greater volume or that have significant diffusion barriers such as cuticles or thick cell walls, one can extend the time to 3 hours simply by putting a lid on the box.
Kit features platform shaker, ice chest, heater block, temperature probe, cryo tubes, and data logger.
Specifications for Product Family
Specifications
  • Catalog No:
  • 103306-432
  • 103307-138
  • Model:
  • 34500-S
  • 34500
Specifications
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Freeze substitution kit without shaker
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $3,404.82
    $3,404.82
    Log in for availability
    Kit of 1
    Kit of 1 - $3,404.82
    $3,404.82
    Each
       $3,404.82
    Kit of 1
       $3,404.82
    Supplier Number: 34500-S
    Specifications:
    More
    Specifications
    Cat. No.
    103306-432
    Model
    34500-S
  • Freeze substitution kit with shaker
    Log in for availability
    Log in for availability
    Each
    Each - $5,445.64
    $5,445.64
    Each
       $5,445.64
    Supplier Number: 34500
    Specifications:
    More
    Specifications
    Cat. No.
    103307-138
    Model
    34500