Labels 'Quality Control'
Labels 'Quality Control'
Quality control label, removable, 22.2×41.4 mm (0.88×1.63")
Catalog # 36471-908
Supplier:   Veriad
Supplier #:  ULLR138
Labels 'Quality Control'
Catalog # 36471-908
Supplier:  Veriad
Labels 'Quality Control'
Quality control label, removable, 22.2×41.4 mm (0.88×1.63")
Price
$84.30
Log in for availability
Pack of 560
Product Details & Documents
Quality control label maintains strict adherence to quality standards and are removable.
Specifications
  • Désignation:
    Quality control label, removable
  • Dimensions:
    22.2×41.4 mm (0.88×1.63")