Microbead Bag, Electron Microscopy Sciences | Avantor
Microbead Bag, Electron Microscopy Sciences
Drawstring bead bag for Thermal microbead 5 L
Catalog # 103302-322
Fournisseur #:  68605-20
Microbead Bag, Electron Microscopy Sciences
Catalog # 103302-322
Microbead Bag, Electron Microscopy Sciences
Microbead Bag, Electron Microscopy Sciences
Drawstring bead bag for Thermal microbead 5 L
Price
$44.12
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For using and storing thermal microbeads.
  • Designed to fit ice buckets
  • Use on top of cool bricks for prolonged cooling
Nylon mesh bag with drawstring closure for storing, cleaning, using, and transporting thermal microbeads. The bag is designed to store microbead in the freezer until ready for use. The bags fits into the 5 L ice bucket.
Spécifications
  • Désignation:
    Drawstring bead bag for Thermal microbead 5 L