À propos de cet article
These precision tweezers from Dumont made of high quality steels with perfect finish suits all application needs.
These tweezers have an anti-magnetic WA1 handle with replaceable 4-ESD tips. Tips attach with screws. Length is 130 mm.
Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as Inox.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.