Multi-Well Embedding Mold, Electron Microscopy Sciences
Multi-Well Embedding Mold, Electron Microscopy Sciences
Multi-Well Embedding Mold, Electron Microscopy Sciences
Certificates
Multi-Well Embedding Mold, Electron Microscopy Sciences
Multi-Well Embedding Mold, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Clear, white or blue silicone mold for embedding.
This unique silicone mold has 2 different cavity sizes to meet all embedding needs. 2 troughs: 1 L x 1/8 W x 1/8" D. 14 cavities: 1/4 L x 1/2 W x 1/8" D. 1 trough: 23/4 L x 3/32 W x 1/8" D. 10 cavities: 3/8 L x 3/4 W x 1/8" D. Each cavity has an alpha-numeric marking, A1-A7, B1-B7, C1-C5, D1-D5. Overall mold measures: 43/4 L x 3 W x 1/4" H (121 x 76 x 6 mm).
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Multi-Well Embedding Mold, Blue
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $52.74
    $52.74
    Each
       $52.74
    Supplier Number: 70914
    Spécifications:
    Marketsource
    Marketsource
  • Multi-Well Embedding Mold, Clear
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $31.77
    $31.77
    Each
       $31.77
    Supplier Number: 70912
    Spécifications:
    Marketsource
    Marketsource
  • Multi-Well Embedding Mold, White
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $37.27
    $37.27
    Each
       $37.27
    Supplier Number: 70913