À propos de cet article
EMS Shield with DAPI is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.
- Does not contain phenylenediamine, which destroys immunofluorescence of Cy dyes, RP-E, PC and APC
- Fortified with DAPI a counter-stain for DNA
- Used in situ hybridization techniques or other methods where fluorescence of DNA staining is required
- RNA is also stained with DAPI
EMS Shield with DAPI is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. This unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Cy2, Cy3, Cy5, Alexa fluoro 488, Alexa fluoro 594, Green fluorescent protein (GFP), tetramethyly rhodamine, Redox. Phycoerythrin (RP-E), Phycocyanin (PC), and Allophycocyanin (APC). Fluorescence is retained during prolonged storage at 4°C in the dark. This medium does not contain phenylenediamine, which destroys immunofluorescence of Cy dyes, RP-E, PC and APC.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Spécifications
- Storage temperature:2-8°C. Protect from light. DO NOT FREEZE.