À propos de cet article
Gun Shot Residue Kit for Scanning Electron Microscopy.
- Powder-free nitrile gloves
- Evidence label
- Chain of custody label on box
- Tamper-evident cardboard transport box (41/2 W x 33/4 D x 3" H)
- Tamper evident seal
The certified SEM pin stubs have very low amounts of Pb, Sb and Ba, not detectable by SEM/EDX. The caps securely hold the GSR sample disc for easy GSR collection. Pre-labeled discs and tubes are both ready-for-sampling. 3-Disc GSR Field Kit has two sampling discs plus a control disc to enhance certainty of the analysis. 4-Disc GSR Field Kit is useful for sampling palm and back separately on both hands. 5-Disc GSR Field Kit is compatible with FBI requirements, with four discs plus a control disc.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.