Thumb Dressing Forceps, Electron Microscopy Sciences
Thumb Dressing Forceps, Electron Microscopy Sciences
Thumb Dressing Forceps, Electron Microscopy Sciences
Thumb Dressing Forceps, Electron Microscopy Sciences
Thumb Dressing Forceps, Electron Microscopy Sciences
These conventional forceps are very popular and are available in a variety of length, tip, and handle styles.
  • Made of premium grade, hardened stainless steel
  • Feature a protective coating for extra durability and protection from rust and corrosion
  • Satin-matte finish reduces eye-fatiguing glare
Product Family Options
  • Désignation
    Overall Length
    Tip Configuration
  • Fine grip forceps
    140 mm
    Serrated tips
    Each
    Each - $125.78
    $125.78
    Each
       $125.78
     : 77937-23
  • Broad grip forceps
    152 mm
    Smooth tips (2 mm)
    Each
    Each - $148.52
    $148.52
    Each
       $148.52
     : 77937-24
  • 305 mm
    Serrated tips
     : -