Mini SEM Clamp, 12.7 mm, Electron Microscopy Sciences
Mini SEM Clamp, 12.7 mm, Electron Microscopy Sciences
Mini SEM Clamp, 12.7 mm
Catalog # 103302-376
Supplier #:  75954-01
Mini SEM Clamp, 12.7 mm, Electron Microscopy Sciences
Catalog # 103302-376
Mini SEM Clamp, 12.7 mm, Electron Microscopy Sciences
Mini SEM Clamp, 12.7 mm
Price
$69.91
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Clamp holder used for flat or thin specimens for Scanning Electron Microscopy.
  • Uses standard 12.7 mm SEM pin stubs
  • Vacuum grade aluminum with brass screws
  • Use for silicon chips, foil, wires, and small tubes
Suitable for holding flat or thin specimens such as silicon chips, foil, wires, and small tubes. Made of Vacuum-grade aluminum with brass screw.
Spécifications
  • Désignation:
    Mini SEM Clamp
  • Opening Size:
    12.7 mm