Earth-Friendly Wide Handle Sampling Spoons, Electron Microscopy Sciences
Earth-Friendly Wide Handle Sampling Spoons, Electron Microscopy Sciences
Earth-Friendly Wide Handle Sampling Spoons, Electron Microscopy Sciences
Earth-Friendly Wide Handle Sampling Spoons, Electron Microscopy Sciences
Retrieve small, specific volume samples of powders or liquids.
  • Wide handles
  • Unique flat bottom shape
  • Dual use as weighing boats
  • Non-sterile
  • Translucent
Long-handled spoons are ideal for acquiring samples from bottles and jars and helping to avoid contact with sample materials. A flat edge on the rim of each bowl allows fast leveling to collect and measure small amounts with accuracy.
Product Family Options
  • Désignation
    Longueur
    Capacité
  • Sampling Spoon
    17.8 cm (7")
    1.25 ml
    Each
    Each - $42.61
    $42.61
    Pack of 10
    Pack of 10 - $33.09
    $33.09
    Each
       $42.61
    Pack of 10
       $33.09
     : 72134
    Marketsource
    Marketsource
  • Sampling Spoon
    17.8 cm (7")
    2.5 ml
    Each
    Each - $42.61
    $42.61
    Pack of 10
    Pack of 10 - $33.09
    $33.09
    Each
       $42.61
    Pack of 10
       $33.09
     : 72136
    Marketsource
    Marketsource
  • Sampling Spoon
    17.8 cm (7")
    5 ml
    Each
    Each - $43.63
    $43.63
    Each
       $43.63
     : 72137
  • Sampling Spoon
    17.8 cm (7")
    8 ml
    Each
    Each - $43.63
    $43.63
    Each
       $43.63
     : 72138
  • Sampling Spoon
    17.8 cm (7")
    20 ml
    Each
    Each - $43.63
    $43.63
    Each
       $43.63
     : 72139