Maxifoam® Premium™ Seamless Knit Gloves, Protective Industrial Products
Supplier: PIP
Certificates
About this item
Designed for general purpose handling in oily and dirty conditions.
- Knit Wrist helps prevent dirt and debris from entering the glove
- 100% Silicone-free
- Light coating gives improved flexibility and dexterity
The surface of the glove designed to 'channel' oil away to give a better grip. With applications in general assembly, micro-engineering, automotive components, painting, horticulture. Recommend washing gloves in tempered water not exceeding 104 °F or 40 °C. A mild commercial laundry soap or detergent should be used.
These Seamless knit nylon gloves feature a nitrile coated micro foam grip on the palm, fingers.
Certifications: Made from component materials which comply with Federal regulations for food contact. 21 CFR, 170-199
Ordering information: This style is also available tri-folded and shrink-wrapped for carefree dispensing through vending machines (34-800V)
Specifications
- Catalog No:
- 20002-390
- 20002-388
- 75814-420
- 20002-386
- 75814-422
- 20002-384
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free